Learn More
Thermo Scientific™ Phadia™ SS-B/La Wells for Connective Tissues Diseases
Catalog No. 14550401P Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
4 x 12 Tests
Catalog No. Quantity
14-550-401P 4 x 12 Tests
1 options
This item is not returnable. View return policy
Catalog No. 14-550-401P Supplier Thermo Scientific™ Supplier No. 14550401
Add to Cart
Add to Cart
This item is not returnable. View return policy
If present in the patient's specimen, antibodies to La bind to their specific antigen.After washing away non-bound antibodies, enzyme-labeled antibodies against human IgG antibodies (EliA IgG Conjugate) are added to form an antibody-conjugate complex. After incubation, non-bound conjugate is washed away and the bound complex is incubated with a Development Solution.After stopping the reaction, the fluorescence in the reaction mixture is measured. To evaluate test results, the response for patient samples is compared directly to the response for calibrators.
  • The EliA La Wells are coated with human recombinant SS-B/La protein
  • The higher the response value, the more specific IgG is present in the specimen.
TRUSTED_SUSTAINABILITY
Specifications
Type SS-B/La Well
For Use With (Application) Connective tissue diseases assay
Quantity 4 x 12 Tests