Learn More
Thermo Scientific™ Phadia™ Tissue Transglutaminase Antibodies for Celiac, Inflammatory Bowel and Other Gastrointestinal Diseases
Catalog No. 14551801P Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
EliA™ Celikey IgA well
EliA™ Celikey IgG well
Quantity:
2 x 12 Tests
4 x 12 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
14-551-801P EliA™ Celikey IgG well 2 x 12 Tests
14-551-701P EliA™ Celikey IgA well 4 x 12 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-551-801P Supplier Thermo Scientific™ Supplier No. 14551801
Add to Cart
Add to Cart
This item is not returnable. View return policy
If present in the patient's specimen, antibodies to tTG bind to their specific antigen. After washing away non-bound antibodies, enzyme-labeled antibodies against human IgA and IgG antibodies, respectively, are added to form an antibody-conjugate complex. After incubation, non-bound conjugate is washed away and the bound complex is incubated with a Development Solution. After stopping the reaction, the fluorescence in the reaction mixture is measured. To evaluate test results, the response for patient samples is compared directly to the response for calibrators.
  • The EliA Celikey Wells are coated with human recombinant tissue transglutaminase (tTG)
  • The higher the response value, the more specific antibodies are present in the specimen.
TRUSTED_SUSTAINABILITY
Specifications
Type EliA™ Celikey IgG well
For Use With (Application) Gastrointestinal diseases assay
Quantity 2 x 12 Tests