Learn More
AMID BIOSCIENCES ER2738 Electrocompetent E. coli Cells, Phage Display, 10 x 0.05 mL
SDP
Catalog No. NC2137275
Encompass
ER2738 electrocompetent E. coli cells are supplied in a 10 x 0.05 mL format for phage display library screening and peptide display workflows. The strain is optimized for high efficiency transformation and M13 phage propagation. Applications include phage display, peptide display libraries, M13 phage propagation, high-efficiency library transformation. Technical specifications are included below.- Quantity - 10 x 0.05 mL (20 reactions)- Organism - E. coli- Storage - -80 C- Cell Strain / Host - ER2738- Cell Format - 10 x 0.05 mL electrocompetent cells- Transformation Efficiency - 1.00E+10This product is for research use only.
This item is not returnable. View return policy
Catalog No. NC2137275 Supplier AMID BIOSCIENCES Supplier No. EREC201
ER2738 electrocompetent E. coli cells are supplied in a 10 x 0.05 mL format for phage display library screening and peptide display workflows. The strain is optimized for high efficiency transformation and M13 phage propagation. Applications include phage display, peptide display libraries, M13 phage propagation, high-efficiency library transformation. Technical specifications are included below.- Quantity - 10 x 0.05 mL (20 reactions)- Organism - E. coli- Storage - -80 C- Cell Strain / Host - ER2738- Cell Format - 10 x 0.05 mL electrocompetent cells- Transformation Efficiency - 1.00E+10This product is for research use only.