Learn More
Boca Scientific Inc Phage T7 DNA, 250ug
Catalog No. NC1894529
Encompass
Bacteriophage T7 DNA is isolated from an infected E. coli strain. Phage T7 DNA is linear with a length of 39936 bp. T7 DNA is the most popular alternative to Lambda DNA substrate for the restriction endonucleases.
Catalog No. NC1894529 Supplier Boca Scientific Inc Supplier No. 310025
Bacteriophage T7 DNA is isolated from an infected E. coli strain. Phage T7 DNA is linear with a length of 39936 bp. T7 DNA is the most popular alternative to Lambda DNA substrate for the restriction endonucleases.