Learn More
Boca Scientific Inc Phage T7 DNA, 50ug
Catalog No. NC1143564
Encompass
Bacteriophage T7 DNA is isolated from infected E.coli strain. Phage T7 DNA is linear with a length of 39936 bp. This DNA is the most popular alternative to Lambda DNA substrate for the restriction endonucleases.
Storage Condition: Store at 18 to 22°C.
Catalog No. NC1143564 Supplier Boca Scientific Inc Supplier No. 310005
Bacteriophage T7 DNA is isolated from infected E.coli strain. Phage T7 DNA is linear with a length of 39936 bp. This DNA is the most popular alternative to Lambda DNA substrate for the restriction endonucleases.
Storage Condition: Store at 18 to 22°C.