Learn More
Abcam Phagocytosis Assay Kit-Green E.coli.Detection method: Fluorescent.Platform: Microplate reader, Fluor. microscope, Flow cyt.
Catalog No. NC2203976
Encompass_Preferred
Phagocytosis Assay Kit-Green E.coli.Detection method: Fluorescent.Platform: Microplate reader, Fluor. microscope, Flow cyt.
The product is subject to the following: Abcam Restricted Use Statement
This item is not returnable. View return policy
Catalog No. NC2203976 Supplier Abcam Supplier No. AB235900100TEST
Phagocytosis Assay Kit-Green E.coli.Detection method: Fluorescent.Platform: Microplate reader, Fluor. microscope, Flow cyt.
The product is subject to the following: Abcam Restricted Use Statement