Learn More
Cayman Chemical PhagocytosIs asy KItIgG-D 1ea
SDP
Catalog No. 502896396
Encompass_Preferred
Cayman’s Phagocytosis Assay Kit (IgG-DyLight™ 405) employs latex beads coated with fluorescently-labeled rabbit IgG as a probe for the measurement of the phagocytic process in vitro. The engulfed fluorescent beads can be detected using a fluorescence microscope, allowing kinetic studies of phagocytosis at the single-cell level. In addition, the flow cytometric readout provides the advantage of visualizing perturbations in phagocytosis on the population level and, when combined with antibody staining, of specific cell types within complex populations. This kit provides enough Latex Beads-Rabbit IgG-DyLight™ 405 Complex for up to 500 samples.
Catalog No. 50-289-6396 Supplier Cayman Chemical Supplier No. 6014801 EA
Cayman’s Phagocytosis Assay Kit (IgG-DyLight™ 405) employs latex beads coated with fluorescently-labeled rabbit IgG as a probe for the measurement of the phagocytic process in vitro. The engulfed fluorescent beads can be detected using a fluorescence microscope, allowing kinetic studies of phagocytosis at the single-cell level. In addition, the flow cytometric readout provides the advantage of visualizing perturbations in phagocytosis on the population level and, when combined with antibody staining, of specific cell types within complex populations. This kit provides enough Latex Beads-Rabbit IgG-DyLight™ 405 Complex for up to 500 samples.