Learn More
Cayman Chemical Phagocytosis Assay Kit (IgG PE), 1 ea
SDP
Catalog No. 502897043
Encompass_Preferred
Cayman’s Phagocytosis Assay Kit (IgG PE) employs latex beads coated with fluorescently-labeled rabbit IgG as a probe for the measurement of the phagocytic process in vitro. The engulfed fluorescent beads can be detected using a fluorescence microscope, allowing kinetic studies of phagocytosis at the single-cell level. In addition, the flow cytometric readout provides the advantage of visualizing perturbations in phagocytosis on the population level and, when combined with antibody staining, of specific cell types within complex populations. This kit provides enough Latex Beads-Rabbit IgG-PE Complex for up to 750 samples.
Catalog No. 50-289-7043 Supplier Cayman Chemical Supplier No. 6005401 EA
Cayman’s Phagocytosis Assay Kit (IgG PE) employs latex beads coated with fluorescently-labeled rabbit IgG as a probe for the measurement of the phagocytic process in vitro. The engulfed fluorescent beads can be detected using a fluorescence microscope, allowing kinetic studies of phagocytosis at the single-cell level. In addition, the flow cytometric readout provides the advantage of visualizing perturbations in phagocytosis on the population level and, when combined with antibody staining, of specific cell types within complex populations. This kit provides enough Latex Beads-Rabbit IgG-PE Complex for up to 750 samples.