Learn More
Medchemexpress LLC HY-P0028 1mg Medchemexpress, Phalloidin CAS:17466-45-4 Purity:>98%
SDP
Catalog No. 502083015
Encompass_Preferred
Medchemexpress, HY-P0028 1mg Phalloidin CAS:17466-45-4 Phalloidin is a mushroom-derived toxin which can be used to label F-actin of the cytoskeleton with fluorochrome(λex=495 nm, λem=520 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3015 Supplier Medchemexpress LLC Supplier No. HYP00281MG
Medchemexpress, HY-P0028 1mg Phalloidin CAS:17466-45-4 Phalloidin is a mushroom-derived toxin which can be used to label F-actin of the cytoskeleton with fluorochrome(λex=495 nm, λem=520 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.