Learn More
Biotium Phalloidin, CF350, 50 U
Catalog No. 501964076
Encompass_Preferred
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. UV-excitable, blue fluorescent CF350 has excitation/emission at 347/448 nm.
Catalog No. 50-196-4076 Supplier Biotium Supplier No. 00049T
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. UV-excitable, blue fluorescent CF350 has excitation/emission at 347/448 nm.