Learn More
Biotium Phalloidin, CF405M, 300 U
Catalog No. 501964062
Encompass_Preferred
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Blue fluorescent CF405M has excitation/emission at 408/452 nm.
Catalog No. 50-196-4062 Supplier Biotium Supplier No. 00034
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Blue fluorescent CF405M has excitation/emission at 408/452 nm.