Learn More
Biotium Phalloidin, CF488A, 300 U
Catalog No. 501964066
Encompass_Preferred
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Freen fluorescent CF488A has excitation/emission at 490/515 nm.
Catalog No. 50-196-4066 Supplier Biotium Supplier No. 00042
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Freen fluorescent CF488A has excitation/emission at 490/515 nm.