Learn More
Biotium Phalloidin, CF568, 300 U
Catalog No. 501964070
Encompass_Preferred
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Red fluorescent CF568 has excitation/emission at 562/583 nm.
Catalog No. 50-196-4070 Supplier Biotium Supplier No. 00044
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Red fluorescent CF568 has excitation/emission at 562/583 nm.