Learn More
Biotium Phalloidin, CF640R, 50 U
Catalog No. 501964078
Encompass_Preferred
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF640R has excitation/emission at 642/662 nm.
Catalog No. 50-196-4078 Supplier Biotium Supplier No. 00050T
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF640R has excitation/emission at 642/662 nm.