Learn More
Biotium Phalloidin, CF647, 300 U
Catalog No. 501964065
Encompass_Preferred
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF647 has excitation/emission at 650/665 nm.
Catalog No. 50-196-4065 Supplier Biotium Supplier No. 00041
Phalloidin conjugates are actin filament stains for fixed and permeabilized cells. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Far-red fluorescent CF647 has excitation/emission at 650/665 nm.