Learn More
Sigma Aldrich Fine Chemicals Biosciences Phalloidin fluorescein Iso1MG
Catalog No. 502904563
Encompass
Phalloidin is a phallotoxin produced by death cap mushroom Amanita phalloides. It is a cyclic peptide which interacts with actin and this was first identified in phalloidin-poisoned rats. It is a heptapeptide cyclic in nature with a crosslink between tryptophan at position 6 and cysteine at position 3. The side chain of amino acid 7 (I3-δ-dihydroxyleucine) in phalloidin is accessible to modifications through which fluorescently labelled phalloidin compounds can be produced.
Catalog No. 50-290-4563 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P5282.1MG
Phalloidin is a phallotoxin produced by death cap mushroom Amanita phalloides. It is a cyclic peptide which interacts with actin and this was first identified in phalloidin-poisoned rats. It is a heptapeptide cyclic in nature with a crosslink between tryptophan at position 6 and cysteine at position 3. The side chain of amino acid 7 (I3-δ-dihydroxyleucine) in phalloidin is accessible to modifications through which fluorescently labelled phalloidin compounds can be produced.