Learn More
Aat Bioquest Phalloidin-iFluor 350 Conjugate (300 Tests)
SDP
Catalog No. 502250142
Encompass_Preferred
This blue fluorescent phalloidin conjugate equivalent to Alexa Fluor 350-labeled phalloidin selectively binds to F-actins. This catalog is for 300 Tests.
This item is not returnable. View return policy
Catalog No. 50-225-0142 Supplier Aat Bioquest Supplier No. 23110
This blue fluorescent phalloidin conjugate equivalent to Alexa Fluor 350-labeled phalloidin selectively binds to F-actins. This catalog is for 300 Tests.