Learn More
Aat Bioquest Phalloidin-iFluor 405 Conjugate (300 Tests)
SDP
Supplier:  Aat Bioquest 23111
Encompass_Preferred
This blue fluorescent phalloidin conjugate equivalent to Alexa Fluor 405-labeled phalloidin selectively binds to F-actins. This catalog is for 300 Tests.
Catalog No. 50-225-0143