Learn More
Aat Bioquest Phalloidin-Tetramethylrhodamine Conjugate (300 Tests)
SDP
Catalog No. 502250140
Encompass_Preferred
This orange fluorescent phalloidin conjugate equivalent to TRITC-labeled phalloidin selectively binds to F-actins with much higher photostability than the fluorescein-phalloidin conjugates. This catalog is for 300 Tests.
This item is not returnable. View return policy
Catalog No. 50-225-0140 Supplier Aat Bioquest Supplier No. 23102
This orange fluorescent phalloidin conjugate equivalent to TRITC-labeled phalloidin selectively binds to F-actins with much higher photostability than the fluorescein-phalloidin conjugates. This catalog is for 300 Tests.