Learn More
Cayman Chemical PhaloIdInIFluor488 Conjug
SDP
Catalog No. 502984620
Encompass_Preferred
A fluorogenic dye used to label F-actin; ex/em spectra of 493/517 nm, respectively; can be used to label formaldehyde-fixed tissue, cells, and cell-free preparations for single- and multi-labeling purposes
Catalog No. 50-298-4620 Supplier Cayman Chemical Supplier No. 20549300 TEST
A fluorogenic dye used to label F-actin; ex/em spectra of 493/517 nm, respectively; can be used to label formaldehyde-fixed tissue, cells, and cell-free preparations for single- and multi-labeling purposes