Learn More
Cayman Chemical Phalloidin-iFluor™ 532 Conjugate, Purity: ≥90%, 300 test
SDP
Catalog No. 502982821
Encompass_Preferred
A fluorogenic dye used to label F-actin; ex/em spectra of 532/556 nm, respectively; can be used to label formaldehyde-fixed tissue, cells, and cell-free preparations for single- and multi-labeling purposes
Catalog No. 50-298-2821 Supplier Cayman Chemical Supplier No. 20551300 TEST
A fluorogenic dye used to label F-actin; ex/em spectra of 532/556 nm, respectively; can be used to label formaldehyde-fixed tissue, cells, and cell-free preparations for single- and multi-labeling purposes