Learn More
Cayman Chemical Phalloidin-iFluor™ 555 Conjugate, Purity: ≥90%, 300 test
SDP
Catalog No. 502984744
Encompass_Preferred
A fluorogenic dye used to label F-actin; ex/em spectra of 556/574 nm, respectively; can be used to label formaldehyde-fixed tissue, cells, and cell-free preparations for single- and multi-labeling purposes
Catalog No. 50-298-4744 Supplier Cayman Chemical Supplier No. 20552300 TEST
A fluorogenic dye used to label F-actin; ex/em spectra of 556/574 nm, respectively; can be used to label formaldehyde-fixed tissue, cells, and cell-free preparations for single- and multi-labeling purposes