Learn More
Cayman Chemical Phalloidin-iFluor™ 594 Conjugate, Purity: ≥90%, 300 test
SDP
Catalog No. 502982074
Encompass_Preferred
A fluorogenic dye used to label F-actin; ex/em spectra of 590/618 nm, respectively; can be used to label formaldehyde-fixed tissue, cells, and cell-free preparations for single- and multi-labeling purposes
Catalog No. 50-298-2074 Supplier Cayman Chemical Supplier No. 20553300 TEST
A fluorogenic dye used to label F-actin; ex/em spectra of 590/618 nm, respectively; can be used to label formaldehyde-fixed tissue, cells, and cell-free preparations for single- and multi-labeling purposes