Learn More
Cayman Chemical Phalloidin-iFluor™ 633 Conjugate, Purity: ≥90%, 300 test
SDP
Catalog No. 502981862
Encompass_Preferred
A fluorogenic dye used to label F-actin; ex/em spectra of 634/649 nm, respectively; can be used to label formaldehyde-fixed tissue, cells, and cell-free preparations for single- and multi-labeling purposes
Catalog No. 50-298-1862 Supplier Cayman Chemical Supplier No. 20554300 TEST
A fluorogenic dye used to label F-actin; ex/em spectra of 634/649 nm, respectively; can be used to label formaldehyde-fixed tissue, cells, and cell-free preparations for single- and multi-labeling purposes