Learn More
3B Scientific™ Phanerogamae Elementary Slide Set
SDPSCI_ED
Catalog No. S95048L Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
25
Catalog No. Quantity
S95048L 25
1 options
This item is not returnable. View return policy
Catalog No. S95048L Supplier 3B Scientific™ Supplier No. W13428
Add to Cart
Add to Cart
This item is not returnable. View return policy
25 microscope slides
  • 1(c). Simple plant cells, epidermis of Allium w.m.
  • 2(d). Cell division (mitosis) all stages, in Allium root tips l.s.
  • 3(c). Starch grains, t.s. of potato tuber
  • 4(c). Cork cells, t.s. of bark of Quercus
  • 5(d). Stone cells, t.s. of fruit of pear
  • 6(d). Root hairs on root tip
  • 7(c). Zea mays, corn, typical monocot root t.s.
  • 8(c). Ranunculus, buttercup, typical dicot root t.s.
  • 9(c). Zea mays, corn, monocot stem t.s.
  • 10(c). Triticum, wheat, gramineous stem t.s.
  • 11(c). Aristolochia, birthwort, one year stem t.s.
  • 12(c). Aristolochia, older stem t.s.
  • 13(d). Cucurbita, pumpkin, stem with bundles and sieve tubes l.s.
  • 14(c). Sambucus, elderberry, stem with lenticels t.s.
  • 15(c). Tulipa, tulip, leaf epidermis with stomata w.m.
  • 16(c). Zea mays, corn, leaf t.s., monocot gramineous leaf
  • 17(c). Syringa, lilac, leaf t.s., dicot leaf
  • 18(c). Fagus, beech, leaf bud t.s. shows leaf origin
  • 19(d). Lilium, lily, flower bud t.s. shows flower diagram
  • 20(d). Lilium, anthers t.s. shows pollen chambers and pollen grains
  • 21(d). Lilium, ovary t.s. with embryosac
  • 22(e). Lilium, stigma with pollen and pollen tubes l.s.
  • 23(c). Pinus, pine, leaf (needle) t.s.
  • 24(d). Triticum, wheat, grain (semen) t.s. with embryo and endosperm
  • 25(d). Capsella, shepherd's purse, l.s. of embryos in situ
Specifications
Type Phanerogamae, Elementary
Quantity 25