Learn More
3B Scientific™ Phanerogamae Supplementary Slide Set
SDPSCI_ED
Catalog No. S95049L Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
50
Catalog No. Quantity
S95049L 50
1 options
Catalog No. S95049L Supplier 3B Scientific™ Supplier No. W13429
Add to Cart
Add to Cart
50 microscope slides
  • 1(d). Stem apex and meristematic tissue, Asparagus l.s.
  • 2(c). Aleurone grains, t.s. of Ricinus endosperm
  • 3(d). Fat in sec. through endosperm of Corylus
  • 4(c). Lysigenous oil glands, t.s. rind of Citrus fruit
  • 5(d). Inulin crystals, t.s. of Dahlia tuber
  • 6(b). Calcium oxalate crystals, in w.m. of onion dry shell
  • 7(b). Wood cells, macerated
  • 8(c). Lactiferous vessels, l.s. stem of Euphorbia (spurge)
  • 9(d). Chloroplasts in leaf of Elodea w.m.
  • 10(b). Branched leaf hairs, isolated from Verbascum (mullein)
  • 11(d). Reserve cellulose, t.s. of Phoenix (date) seed
  • 12(c). Rheum, rhubarb, root with crystals t.s.
  • 13(c). Dendrobium, orchid, aerial root with velamen t.s.
  • 14(c). Pinus, pine, older woody root t.s.
  • 15(c). Smilax, root with thickened endodermis t.s.
  • 16(d). Lupinus, lupin, root nodules with nitrogen fixing bacteria t.s.
  • 17(c). Quercus, oak, older woody root t.s.
  • 18(c). Daucus, carrot, storage root t.s.
  • 19(c). Pinus, pine, older woody stem t.s.
  • 20(c). Zea mays, corn, stem with bundles l.s.
  • 21(c). Elodea, waterweed, aquatic stem with primitive bundle t.s.
  • 22(c). Juncus, bulrush, stem with stellate cells t.s.
  • 23(c). Pelargonium, geranium, young stem of an annual plant t.s.
  • 24(d). Tilia, lime, older woody stem t.s. and l.s.
  • 25(c). Acorus calamus, sweet flag, rhizome t.s.
  • 26(d). Pinus, pine, three sections of wood
  • 27(d). Fagus, beech, three sections of wood
  • 28(c). Bryonia, stem with sieve plates t.s.
  • 29(c). Ribes, currant, stem with phellogen t.s.
  • 30(c). Helianthus, sunflower, typical dicot stem t.s.
  • 31(c). Salvia, sage, square stem with collenchyma t.s.
  • 32(c). Nymphaea, water lily, floating leaf t.s.
  • 33(c). Dionaea, Venus fly trap, leaf with digestive glands t.s.
  • 34(d). Fagus, beech, sun and shadow leaves on one slide t.s.
  • 35(c). Pinguicula, butterwort, leaf with glandular hairs t.s.
  • 36(c). Nerium, oleander, xeromorphe leaf with sunken stomata t.s.
  • 37(d). Drosera, sundew, leaf with glandular hairs w.m.
  • 38(d). Urtica, stinging nettle, leaf with stinging hairs
  • 39(c). Utricularia, bladderwort, w.m. of bladders
  • 40(d). Pinus, pine, male cone with pollen l.s.
  • 41(d). Pinus, young female cone with ovules l.s.
  • 42(f). Pinus, ovule with archegonia l.s.
  • 43(e). Pinus, mature embryo with endosperm t.s.
  • 44(b). Pinus, pollen grains with wings w.m.
  • 45(f). Lilium, lily, young anthers showing meiosis of pollen mother cells
  • 46(d). Tulipa, tulip, ovary t.s. showing arrangement of ovules
  • 47(d). Taraxacum, dandelion, composite flower l.s.
  • 48(d). Papaver, poppy, flower t.s. shows floral diagram
  • 49(d). Phaseolus, bean, pod showing pericarp and seed t.s.
  • 50(d). Lycopersicum, tomato, young fruit t.s.
Specifications
Type Phanerogamae
Quantity 50