Learn More
Proteintech Group Inc Phantom Dye Red 780 Viability Dye
SDP
Supplier:  Proteintech Group Inc PD00002100TESTS
Encompass_Preferred
Phantom Dye Red 780 is a fixable viability dye which reacts with protein amine groups in both dead and live cells. Due to the compromised cell membrane in dead cells the dye can bind to many more proteins inside the cell, resulting in much higher fluorescence and easy differentation of Live/Dead cells. Phantom Dye Red 780 is a water soluble red fluorescent dye excited by the red laser (633 nm) making it an excellent addition to your multicolor panel.
Catalog No. 50-225-8577