Learn More
Proteintech Group Inc Phantom Dye UV 450 Viability Dye
SDP
Supplier:  Proteintech Group Inc PD00003100TESTS
Encompass_Preferred
Phantom Dye UV 450 is a fixable viability dye which reacts with protein amine groups in both dead and live cells. Due to the compromised cell membrane in dead cells the dye can bind to many more proteins inside the cell, resulting in much higher fluorescence and easy differentation of Live/Dead cells. Phantom Dye UV 450 is a water soluble UV fluorescent dye excited by the UV laser (355 nm) making it an excellent addition to your multicolor panel.
Catalog No. 50-225-8578