Learn More
MIGALI SCIENTIFIC Pharm/Vaccine/Lab Freezer 44.9 cu ft
Supplier:  MIGALI SCIENTIFIC EVOX2FPH
Encompass_Preferred
Pharm/Vaccine/Lab Freezer -30 C auto defrost freezer with standard baskets
Catalog No. NC2964403
Add to Cart
Request a Quote