Learn More
MIGALI SCIENTIFIC Pharm/Vaccine/Lab undercounter Freezer
Supplier:  MIGALI SCIENTIFIC EVOXU1FLB
Encompass_Preferred
Pharm/Vaccine/Lab undercounter Freezer -30C Auto Defrost Solid Door 4.2 cu ft
Catalog No. NC2964437