Learn More
MIGALI SCIENTIFIC Pharm/Vaccine/Lab undercounter Freezer
Catalog No. NC2964438
Encompass_Preferred
Pharm/Vaccine/Lab undercounter Freezer -30C Auto Defrost Solid Door 4.2 cu ft with Standard Baskets
Catalog No. NC2964438 Supplier MIGALI SCIENTIFIC Supplier No. EVOXU1FPH
Pharm/Vaccine/Lab undercounter Freezer -30C Auto Defrost Solid Door 4.2 cu ft with Standard Baskets