Learn More
Acute Care Pharmaceuticals Pharma-D™ Sterile Disinfectant ; Virucide, Germicide, Sanitizer 16OZ trigger sprayer
Catalog No. NC0969459
Encompass_Preferred
Pharma-D Surface Disinfectant™ is an EPA registered, validated sterile, one-step germicidial/disinfectant cleaner that is available in 16oz RTU spray bottles. Pharma-D™ is a quaternaryammonium designed to remove surface contaminants and disinfect hard, non-poroussurface areas. It can be used as an additional disinfectant in rotation withPharma-Hol™, in your cleanroom, glove box, or compounding areas. 12 bottles per case, double bagged, COA's available upon request.
This item is not returnable. View return policy
Catalog No. NC0969459 Supplier Acute Care Pharmaceuticals Supplier No. PDQUAT79716
Pharma-D Surface Disinfectant™ is an EPA registered, validated sterile, one-step germicidial/disinfectant cleaner that is available in 16oz RTU spray bottles. Pharma-D™ is a quaternaryammonium designed to remove surface contaminants and disinfect hard, non-poroussurface areas. It can be used as an additional disinfectant in rotation withPharma-Hol™, in your cleanroom, glove box, or compounding areas. 12 bottles per case, double bagged, COA's available upon request.