Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Pharma KingFisher™ Flex 96 Deep-Well Plates

Catalog No. A43075
Change view
Click to view available options
Quantity:
50 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A43075 50 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A43075 Supplier Applied Biosystems™ Supplier No. A43075
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These deep-well plates are for use with the Pharma KingFisher Flex 96 Deep-Well Head Magnetic Particle Processor or MagMAX Express-96 Deep-Well Magnetic Particle Processor in combination with PrepSEQ ResDNA kits and ProteinSEQ kits.

The deep-well configuration differs from the standard configuration in that the volume of sample input is from 100 to 1,000 μL per well compared to 20–200 μL for the standard configuration.

Optimize performance using KingFisher Flex System consumables designed specifically for use with these instruments. KingFisher Flex disposable plastics (tip combs and microplates) are made of polypropylene and are ideal for magnetic particle processing because of their low binding affinity for biomolecules. Their special design enables excellent recovery of magnetic beads.

Specifications

Barcode No Barcode
Content And Storage 50 deep-well 96-well plates
Packaging 50 Plates
Product Line KingFisher Flex
Color Clear
Product Type Microplate
For Use With (Equipment) KingFisher™ Flex
Material Polypropylene
No. of Wells 96
Quantity 50 plates
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.