Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ Pharma KingFisher™ Flex 96 Standard Plates

Catalog No. A43076
Change view
Click to view available options
Quantity:
48 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A43076 48 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A43076 Supplier Applied Biosystems™ Supplier No. A43076
Get an Extra Analytical Consumable with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These standard plates are for use with the Pharma KingFisher Flex 96 Deep-Well Head Magnetic Particle Processor or MagMAX Express-96 Standard Magnetic Particle Processor in combination with PrepSEQ ResDNA kits and ProteinSEQ kits.

These standard plates are for use with the Pharma KingFisher Flex 96 Deep-Well Head Magnetic Particle Processor or MagMAX Express-96 Standard Magnetic Particle Processor in combination with PrepSEQ ResDNA kits and ProteinSEQ kits. The standard configuration differs from the deep-well configuration in that the volume of sample input is from 20–200 μL per well compared to 100–1000 μL per well for the deep-well configuration.

Optimize performance using KingFisher Flex System consumables designed specifically for use with these instruments. KingFisher Flex disposable plastics (tip combs and microplates) are made of polypropylene and are ideal for magnetic particle processing because of their low binding affinity for biomolecules. Their special design enables excellent recovery of magnetic beads.

Specifications

Barcode No Barcode
Content And Storage 48 standard-well 96-well plates
Array 8 x 12
Packaging 48 Plates
Product Line KingFisher Flex
Color Clear
Product Type Microplate
For Use With (Equipment) KingFisher™ Flex
Material Polypropylene
No. of Wells 96
Quantity 48 Plates
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.