Learn More
Pharma Magnetic Stand-96
Catalog No. A31543
Change view
Click to view available options
Quantity:
1 Stand
Catalog No. Quantity
A31543 1 Stand
1 options
Catalog No. A31543 Supplier Thermo Scientific Supplier No. A31543
Add to Cart
Add to Cart
The Magnetic Stand-96 is designed for paramagnetic bead precipitation from standard 96-well, U-bottom microplates and 0.2 mL PCR plates with no additional accessories.
The stand has a clear polyacryl base in a standard SBS 96-well microplate footprint. The use of 24 extremely strong permanent magnets enables easy and fast magnetic separation in as little as 30 seconds. It sits flat on a lab bench as well as in a microplate adapter on robotic workstations.
Specifications
Shipping Condition Room Temperature
Product Line KingFisher™ Flex
Quantity 1 Stand
Type Magnetic Stand
For Research Use Only. Not for use in diagnostic procedures.