Learn More
Foxx Life Sciences EZBio Stainless Steel Pharma Scoop Polished with Steel Handle, 316L, 50mL, 1/EA
Catalog No. NC1795163
Encompass_Preferred
Foxx Life Sciences Stainless Steel Pharma Scoops fulfill or exceed the high safety and regulatory compliance requirements in production and sampling for the pharmaceutical industry. These exceptional products are made of high-quality, corrosion and chemical resistant, 316L Stainless Steel. The hygienic design and mirror finish make them easy to clean with no crevices or sharp edges. They are ideal for all types of work from the laboratory to production and especially for use with powders and granules. These superior products are available in several sizes, ranging from 10mL to 2500mL.Heavy duty, high strength, chemical and corrosion resistant, 316L SSMirror polish on inner and outer surfacesFully welded to remove and crevices and dead spacesEasy to clean hygienic designCorners are radiused in line with GMP guidelines
Catalog No. NC1795163 Supplier Foxx Life Sciences Supplier No. 25C3802FLS
Foxx Life Sciences Stainless Steel Pharma Scoops fulfill or exceed the high safety and regulatory compliance requirements in production and sampling for the pharmaceutical industry. These exceptional products are made of high-quality, corrosion and chemical resistant, 316L Stainless Steel. The hygienic design and mirror finish make them easy to clean with no crevices or sharp edges. They are ideal for all types of work from the laboratory to production and especially for use with powders and granules. These superior products are available in several sizes, ranging from 10mL to 2500mL.Heavy duty, high strength, chemical and corrosion resistant, 316L SSMirror polish on inner and outer surfacesFully welded to remove and crevices and dead spacesEasy to clean hygienic designCorners are radiused in line with GMP guidelines