Learn More
eMolecules​ 947533-45-1 | 2-bromo-5-fluoropyrimidine | Pharmablock | MFCD09835164 | 176.976 | C4H2BrFN2 | 97.000 | Fc1cnc(Br)nc1 | 100mg | 551127796
Catalog No. 5000127623 Shop All eMolecules Inc. Products
Encompass_Preferred
2-bromo-5-fluoropyrimidine | Pharmablock | 947533-45-1 | MFCD09835164 | 176.976 | C4H2BrFN2 | 97.000 | Fc1cnc(Br)nc1 | 100mg | 551127796
Catalog No. 50-001-27623 Supplier eMolecules​ Supplier No. 551127796
2-bromo-5-fluoropyrimidine | Pharmablock | 947533-45-1 | MFCD09835164 | 176.976 | C4H2BrFN2 | 97.000 | Fc1cnc(Br)nc1 | 100mg | 551127796