Learn More
PHCbi TwinGuard ECO Series Pharmaceutical Refrigerator
Catalog No. 502720038
Encompass_Preferred
The PHCbi brand TwinGuard ECO Series MPR-S1201XH-PA Pharmaceutical Refrigerator provides twice the sample protection for double the peace of mind. Two independent cooling circuits work together, both independently and concurrently, to provide a comprehensive solution to potential challenges, ensuring the continuity and safety of your precious contents. The cutting-edge refrigeration system ensures reliable, uniform temperature control. Natural HC refrigerants and state-of-the-art inverter-controlled compressors achieve a significant reduction in power consumption and greater uniformity than predecessor models.Effective capacity: 41.1 cu.ft. (1165 L)Temperature range: 2℃ to 14℃
Catalog No. 50-272-0038 Supplier PHCbi Supplier No. MPRS1201XHPA
Add to Cart
Add to Cart
Request a Quote
The PHCbi brand TwinGuard ECO Series MPR-S1201XH-PA Pharmaceutical Refrigerator provides twice the sample protection for double the peace of mind. Two independent cooling circuits work together, both independently and concurrently, to provide a comprehensive solution to potential challenges, ensuring the continuity and safety of your precious contents. The cutting-edge refrigeration system ensures reliable, uniform temperature control. Natural HC refrigerants and state-of-the-art inverter-controlled compressors achieve a significant reduction in power consumption and greater uniformity than predecessor models.Effective capacity: 41.1 cu.ft. (1165 L)Temperature range: 2℃ to 14℃