Learn More
Perkin Elmer US LLC Pharmaceutical OVI Application Consumables Kit
Catalog No. 502112459
Encompass_Preferred
Kit includes: Elite-1301 30 M x 0.53 mm x 3.0µm column, Zero Dilution Glass Outer Liner, Zero Dilution Glass Inner Liner, Polyimide uncoated deactivated tubing, 5 M x 0.18 mm, 22 mL headspace vials, 19 mm assembled cap and septa PTFE/silicone and 20 mm electronic crimper (120V).
Catalog No. 50-211-2459 Supplier Perkin Elmer US LLC Supplier No. N9300655
Add to Cart
Add to Cart
Request a Quote
Kit includes: Elite-1301 30 M x 0.53 mm x 3.0µm column, Zero Dilution Glass Outer Liner, Zero Dilution Glass Inner Liner, Polyimide uncoated deactivated tubing, 5 M x 0.18 mm, 22 mL headspace vials, 19 mm assembled cap and septa PTFE/silicone and 20 mm electronic crimper (120V).