Learn More
PHCbi Pharmaceutical refrigerator
Catalog No. 502720044
Encompass_Preferred
PHCbi brand 24.2 cu.ft (684 L) Upright pharmacy refrgierator offers a practical solution for storage with ajustable shelves and a wide setpoint range for varying applications. ENERGY STAR CertifiedTemperature Range2 to 23
Catalog No. 50-272-0044 Supplier PHCbi Supplier No. MPR722PA
Add to Cart
Add to Cart
Request a Quote
PHCbi brand 24.2 cu.ft (684 L) Upright pharmacy refrgierator offers a practical solution for storage with ajustable shelves and a wide setpoint range for varying applications. ENERGY STAR CertifiedTemperature Range2 to 23