Learn More
PHCbi 48.2 cu.ft. Pharmacy Refrigerator
Catalog No. 502720046
Encompass_Preferred
The MPR-1412-PA is a large capacity pharmacy refrigerator delivering a stable and reliable environment for managing clinical, pharmaceutical and biomedical storage and processing at uniform temperatures. Easy to use features and design and precise temperature control with fast recovery following door openings, assures safe storage during frequent access and high ambient conditions.Effective capacity: 48.2 cu.ft. (1364 L)Temperature range: 2℃ to 23℃
Catalog No. 50-272-0046 Supplier PHCbi Supplier No. MPR1412PA
Add to Cart
Add to Cart
Request a Quote
The MPR-1412-PA is a large capacity pharmacy refrigerator delivering a stable and reliable environment for managing clinical, pharmaceutical and biomedical storage and processing at uniform temperatures. Easy to use features and design and precise temperature control with fast recovery following door openings, assures safe storage during frequent access and high ambient conditions.Effective capacity: 48.2 cu.ft. (1364 L)Temperature range: 2℃ to 23℃