Learn More
Helix Medical Inc PHARMAFOCUS TUBING
Catalog No. NC0901658
Encompass_Preferred
0.125 x 0.250" Biomedical/pharmaceutical grade standard tubing, made with Wacker Silicones HM-6540 platinum-cured material. The manufacturing process minimizes flow degradation over extended periods. Meets USP Class VI biocompatibility requirements.
Catalog No. NC0901658 Supplier Helix Medical Inc Supplier No. 6041151
0.125 x 0.250" Biomedical/pharmaceutical grade standard tubing, made with Wacker Silicones HM-6540 platinum-cured material. The manufacturing process minimizes flow degradation over extended periods. Meets USP Class VI biocompatibility requirements.