Learn More
MITSUBISHI GAS CHEMICAL 20cc O2 and H2O Absorber
Catalog No. NC2602568
Encompass
PHARMAKEEP KD-20, Packet, 4000 pcs/carton, >20 ml oxygen absorption capacity, >0.15 g moisture absorption capacity, 26 mm x 62 mm
Catalog No. NC2602568 Supplier MITSUBISHI GAS CHEMICAL Supplier No. KD20
PHARMAKEEP KD-20, Packet, 4000 pcs/carton, >20 ml oxygen absorption capacity, >0.15 g moisture absorption capacity, 26 mm x 62 mm