Learn More
Nordson Medical PHARMALOK LRG TUBE CLAMP 100PK
Catalog No. NC2206733
Encompass_Preferred
PharmaLok™ Large Tube Clamp, wall thickness 1/8” (3.2mm) to 1/4”(6.4mm), max OD 2.5” (63.5mm), Glass filled orange nylon 100/pk
This item is not returnable. View return policy
Catalog No. NC2206733 Supplier Nordson Medical Supplier No. CPT110U706
PharmaLok™ Large Tube Clamp, wall thickness 1/8” (3.2mm) to 1/4”(6.4mm), max OD 2.5” (63.5mm), Glass filled orange nylon 100/pk