Learn More
Nordson Medical PharmaLok ZerO Clamp Mini
Catalog No. 503038163
Encompass_Preferred
PharmaLok ZerO Clamp Series Hygienic Union TC Version 5 (12 7 mm) to 75 (19 0 mm) Glass Reinforced White Nylon with Blue Lever
Catalog No. 50-303-8163 Supplier Nordson Medical Supplier No. CPUN075U71
PharmaLok ZerO Clamp Series Hygienic Union TC Version 5 (12 7 mm) to 75 (19 0 mm) Glass Reinforced White Nylon with Blue Lever