Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pharmalytes™ pH range 4.0 - 6.5, MilliporeSigma™ Supelco™

Catalog No. 111015313 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5313 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5313 Supplier MilliporeSigma Supelco Supplier No. P177225ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmalytes™ are ampholytes which are used to create a pH gradient under application of an electric field.

TRUSTED_SUSTAINABILITY

Specifications

CAS 70852-56-1
Content And Storage 2°C to 8°C
Product Line Pharmalyte
Quantity 25 mL
Synonym Ampholytes

Pharmalyte is a registered trademark of GE Healthcare

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.