Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foamtec PharmaMOP™ Bucket

Catalog No. 17184110 Shop All Foamtec International Products
Change view
Click to view available options
Dimensions:
20.5 x 10.5 x 10.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions
17-184-110 20.5 x 10.5 x 10.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-184-110 Supplier Foamtec Supplier No. FT409BL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for critical cleanroom surfaces.

  • Suitable for cleaning ceilings, walls, and floors
  • Simple press-fit connection allows gloved operators to easily attach mop heads
  • Compatible with steam sterilization cycles
  • Mop heads provide quick and efficient removal of disinfectant residues
  • Features a two-direction, 360° swivel mop head
  • Made with abrasion-resistant cleanroom fabric
  • Class 100 processed
  • Provides fiber-free mopping of rough and non-skid floor
  • Sterile
  • Features an ultra-lightweight telescoping handle

Specifications

Product Type Bucket
Material Polypropylene
Autoclavable Autoclavable
Certifications/Compliance ISO 9001:2000, ISO 13485:2003
Dimensions 20.5 x 10.5 x 10.5 in.
Disposable Reusable
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.