Learn More
Foamtec PharmaMOP™ GO Stainless Steel V-Bracket
Catalog No. 17111171 Shop All Foamtec International Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
17-111-171 Stainless Steel
1 options
Catalog No. 17-111-171 Supplier Foamtec Supplier No. FT432
Add to Cart
Add to Cart
For critical cleanroom surfaces allowing the operator to clean ceilings, walls and floors with ease.
  • Utilizes simple press fit connection that enables gloved operators to easily attach mop heads and is compatible with steam sterilization cycles
  • Mop heads enable the quick, efficient cleaning of surfaces stained with disinfectant residues
  • Two direction 360° swivel and mop head
  • Abrasion resistant cleanroom fabric
  • Class 100 processing
  • Fiber-free mopping of rough, non-skid floors
  • Sterile
  • Ultra-lightweight telescoping handle
Specifications
Product Type Bucket Wringer Frame
Material Stainless Steel
Autoclavable Autoclavable
Certifications/Compliance ISO 9001:2000, 13485:2003
Disposable Reusable
Sterility Non-sterile