Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S High-Performance Precision Pump Tubing, PharmaPure, L/S 24; 25 ft
Catalog No. NC9446539
Encompass
Applications: Pharmaceutical and biotech Characteristics: Long life under pressure up to 60 psi (4 bar), excellent biocompatibility, low extractables, very low spallation, nontoxic, nonhemolytic, opaque off-white
Catalog No. NC9446539 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0643524/DEL
Applications: Pharmaceutical and biotech Characteristics: Long life under pressure up to 60 psi (4 bar), excellent biocompatibility, low extractables, very low spallation, nontoxic, nonhemolytic, opaque off-white