Learn More
Accuflow Systems Saint-Gobain PharmaPure Peristaltic Tubing, 1/4 in ID, 3/8 in OD
Catalog No. 502365783
Encompass_Preferred
PharmaPure peristaltic pump tubing is an ultra-pure, low-spallation multilayer thermoplastic elastomer tubing engineered for bioprocessing and laboratory fluid transfer. It provides extended pump life, very low particulate generation, and low gas permeability while maintaining mechanical stability after autoclaving and high-dose gamma irradiation.
  • Low particulate spallation and extended pump life.
  • 1/4 in ID x 3/8 in OD, standard 25 ft length.
  • Proprietary multilayer thermoplastic elastomer construction.
  • Autoclavable and maintains performance after gamma irradiation up to 50 kGy.
  • Meets USP Class VI biological test for plastics; documented biocompatibility.
  • Low gas permeability to protect sensitive media and cultures.
  • Suitable for peristaltic pumps, sterile filling, bioreactors, and single-use systems.
Catalog No. 50-236-5783 Supplier Accuflow Systems Supplier No. AL242017
PharmaPure peristaltic pump tubing is an ultra-pure, low-spallation multilayer thermoplastic elastomer tubing engineered for bioprocessing and laboratory fluid transfer. It provides extended pump life, very low particulate generation, and low gas permeability while maintaining mechanical stability after autoclaving and high-dose gamma irradiation.
  • Low particulate spallation and extended pump life.
  • 1/4 in ID x 3/8 in OD, standard 25 ft length.
  • Proprietary multilayer thermoplastic elastomer construction.
  • Autoclavable and maintains performance after gamma irradiation up to 50 kGy.
  • Meets USP Class VI biological test for plastics; documented biocompatibility.
  • Low gas permeability to protect sensitive media and cultures.
  • Suitable for peristaltic pumps, sterile filling, bioreactors, and single-use systems.